missing translation for 'onlineSavingsMsg'
Learn More

BD Bottled Media Saboraud Dextrose Agar

For cultivation and maintenance of pathogenic and non-pathogenic fungi

$700.95 - $700.95

Specifications

Form Liquid
Manufacturing Origin United States
Fill Volume 500 mL
Formulation Sabouraud Detrose Agar
For Use With (Application) Used for qualitative procedures for cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes
View More Specs

Products 1
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
B4399510
View Documents
BD
299510
No Chloramphenicol
Pack of 10 Each for $700.95
Only null left
 
Description

Description

  • Volume: 500mL
  • With and without Chloramphenicol

Specifications

Specifications

Liquid
500 mL
Used for qualitative procedures for cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes
500 mL
United States
Sabouraud Detrose Agar
Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'