missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Taxo™ P Discs (Vial)

Catalog No. B31047
Change view
Click to view available options
Packaging Type:
6 Vials
Vial
Quantity:
50 Discs
6 x 50 Discs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type Quantity
B31047 6 Vials 6 x 50 Discs
B31046 Vial 50 Discs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31047 Supplier BD Supplier No. 231047
Edge

For presumptive identification of pneumococci, and are intended for use with pure cultures

  • 6mm discs made from high quality absorbent paper
  • Impregnated with approximately 5.0μg of ethylhydrocupreine hydrochloride per disc

Order Info

Vials of 50 discs.

Specifications

Antimicrobial Agents Ethylhydrocupreine hydrochloride (optochin)
Concentration 5 μg
Content And Storage -20°C to +8°C
For Use With (Application) Differentiating Pneumococci
Packaging Type 6 Vials
Quantity 6 x 50 Discs

For in vitro Diagnostic Use.