Learn More
BD BBL™ Taxo™ Differentiation Discs, Novobiocin
For determination of novobiocin resistance in coagulase negative staphylococci
Supplier: BD 231750
Description
- Absorbant paper containing 5μg novobiocin
Specifications
| Novobiocin | |
| -20°C to +8°C | |
| 1 Pack |
| 5 μg | |
| Disc |
Safety and Handling
ShelfLife : 730 Days
For in vitro Diagnostic Use.