missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Taxo™ A Discs (Cartridge)

Catalog No. p-4897816
Change view
Click to view available options
Packaging Type:
10 Cartridges
Quantity:
10 x 50 Discs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type Quantity
B31552 10 Cartridges 10 x 50 Discs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B31552 Supplier BD Supplier No. 231552
Only null left

For presumptive identification of group A beta-hemolytic streptococci based on susceptibility to a low level of bacitracin

  • Impregnated with approximately 0.04 units of bacitracin per disc

Taxo A discs are not for susceptibility testing.

Order Info

Cartridge of 50 discs

Specifications

Antimicrobial Agents Bacitracin
Concentration 0.04U
Content And Storage -20°C to +8°C
For Use With (Application) For differentiating Group A Streptococci
Packaging Type 10 Cartridges
Quantity 10 x 50 Discs

For in vitro Diagnostic Use.