missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Staphyloslide™ Latex Test Kit
Provides a rapid platform for the identification of Staphylococcal isolates, particularly, Staphylococcus aureus
Supplier: BD 240915
Includes: BBL Staphyloslide Test Latex; BBL Staphyloslide Control Latex; Test cards, Mixing sticks
Description
The BD BBL Staphyloslide Latex Test Kit utilizes blue polystyrene latex particles that have been sensitized with fibrinogen and IgG. When Staphylococcal colonies which possess bound coagulase (clumping factor) and/or protein A are mixed with the latex reagent, the latex particles will agglutinate strongly within 20 seconds.
Recommended Storage
Store at 2° to 8°C. Do not freeze.
Specifications
| 2°C to 8°C Do not freeze | |
| S. aureus | |
| BBL Staphyloslide Test Latex, 2 x 2.5mL; BBL Staphyloslide Control Latex, 2.5mL; Test cards, Mixing sticks |
| BBL Staphyloslide Latex Test Kit | |
| Staphylococcus aureus Testing | |
| 100 Tests |