missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Staphyloslide™ Latex Test Kit

Catalog No. B240916
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B240916 500 Tests
B240915 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B240916 Supplier BD Supplier No. 240916

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

BBL Staphyloslide Test Latex; BBL Staphyloslide Control Latex; Test cards, Mixing sticks

Provides a rapid platform for the identification of Staphylococcal isolates, particularly, Staphylococcus aureus

The BD BBL Staphyloslide Latex Test Kit utilizes blue polystyrene latex particles that have been sensitized with fibrinogen and IgG. When Staphylococcal colonies which possess bound coagulase (clumping factor) and/or protein A are mixed with the latex reagent, the latex particles will agglutinate strongly within 20 seconds.

Recommended Storage

Store at 2° to 8°C. Do not freeze.

Specifications

Content And Storage 2°C to 8°C Do not freeze
Description BBL Staphyloslide Latex Test Kit
Detectable Analytes S. aureus
For Use With (Application) Staphylococcus aureus Testing
Includes BBL Staphyloslide Test Latex, 5 x 5mL; BBL Staphyloslide Control Latex, 2 x 6.5mL; Test cards, Mixing sticks
Quantity 500 Tests