Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Oxacillin
Description
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Specifications
| Antimicrobial Agents | Oxacillin |
| Concentration | 1 μg |
| Packaging Type | Disc |
| Quantity | 1/Pk. |