Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Azlocillin,7 AZ-7
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
Supplier: BD 231625
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
- Employs Bauer-Kirby disc diffusion testing method
- Use in self-tamping dispensers
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
- Cartridges of 50 discs each are sold individually or in packs of 10
Specifications
| Azlocillin | |
| 75 μg | |
| Disc | |
| 10 Cartridges |
Safety and Handling
ShelfLife : 730 Days