Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Ampicillin4, AM-10
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
$36.42 - $179.19
Specifications
| Antimicrobial Agents | Ampicillin |
|---|---|
| Concentration | 10 μg |
| Packaging Type | Disc |
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
- Employs Bauer-Kirby disc diffusion testing method
- Use in self-tamping dispensers
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
- Cartridges of 50 discs each are sold individually or in packs of 10
Specifications
| Ampicillin | |
| 10 μg | |
| Disc |