Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Amoxicillin w/Clavulanic Acid3, AmC-30
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
$38.90 - $191.07
Specifications
Antimicrobial Agents Amoxicillin w/ Clavulanic Acid
Concentration 20/10 μg
Packaging Type Disc
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
B31629
Accessories View Documents Promotion Details
BD
231629
10 Cartridges
Case of 10 Each for $191.07
Only null left
B31628
View Documents Promotion Details
BD
231628
1 Cartridge
Each for $38.90
Only null left
Description
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Employs Bauer-Kirby disc diffusion testing method
  • Use in self-tamping dispensers
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
  • Cartridges of 50 discs each are sold individually or in packs of 10
Specifications
Specifications
Amoxicillin w/ Clavulanic Acid
20/10 μg
Disc
Videos
Safety and Handling
Safety and Handling
ShelfLife : 548 Days