Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Piperacillin/Tazobactam60
Catalog No. B231691
Change view
Click to view available options
Quantity:
1 Pack
10 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B231691 1 Pack
B31692 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B231691 Supplier BD Supplier No. 231691
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Antimicrobial Agents Piperacillin, Tazobactam 60
Concentration 100/10 μg
Packaging Type Disc
Quantity 1 Pack
For in vitro diagnostic use