Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Colistin35
Catalog No. B31278
Change view
Click to view available options
Quantity:
1 Pack
10 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31278 10 Pack
B30749 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31278 Supplier BD Supplier No. 231278
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Antimicrobial Agents Colistin35
Concentration 10 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro diagnostic use