Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Colistin35
Catalog No. B30749
Change view
Click to view available options
Quantity:
1 Pack
10 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30749 1 Pack
B31278 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30749 Supplier BD Supplier No. 230749
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Antimicrobial Agents Colistin35
Concentration 10 μg
Packaging Type Disc
Quantity 1 Pack
For in vitro diagnostic use