Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Clindamycin33
Catalog No. B31275
Change view
Click to view available options
Quantity:
1 Pack
10 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31275 10 Pack
B31213 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31275 Supplier BD Supplier No. 231275
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Antimicrobial Agents Clindamycin33
Concentration 2 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro diagnostic use