Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ciprofloxacin31
Catalog No. B31657
Change view
Click to view available options
Quantity:
1 Cartridge (50 Discs/Cartridge)
10 Cartridges (50 Discs/Cartridge)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31657 1 Cartridge (50 Discs/Cartridge)
B31658 10 Cartridges (50 Discs/Cartridge)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31657 Supplier BD Supplier No. 231657
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Antimicrobial Agents Ciprofloxacin31
Concentration 5 μg
Packaging Type Disc
Quantity 1 Cartridge (50 Discs/Cartridge)
For in vitro diagnostic use