Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ceftazidime/Clavulanic Acid
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Supplier: BD 231753
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
| Ceftazidime, Clavulanic Acid | |
| 10 μg | |
| Disc | |
| 1 Pack |
For in vitro diagnostic use