Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefotetan19
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Supplier: BD 231656
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
| Cefotetan19 | |
| 30 μg | |
| Disc | |
| 10 Pack |
Safety and Handling
ShelfLife : 730 Days
For in vitro diagnostic use