Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Aztreonam8
Catalog No. B31640
Change view
Click to view available options
Quantity:
10 Pack
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31640 Each
B31641 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31640 Supplier BD Supplier No. 231640
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Antimicrobial Agents Aztreonam8
Concentration 30 μg
Includes 50 discs/cartridge
Packaging Type Disc
Quantity Each
For in vitro diagnostic use