Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Azithromycin6
Catalog No. B31682
Change view
Click to view available options
Concentration:
15 μg
Catalog No. Concentration
B31682 15 μg
1 options
Catalog No. B31682 Supplier BD Supplier No. 231682
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Antimicrobial Agents Azithromycin6
Concentration 15 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro diagnostic use