Learn More
BD BBL™ Sensi-Disc™ Antibiotic Susceptibility Test Discs Accessories: Sensi-Disc Dispenser
Catalog No. 148908
Change view
Click to view available options
Packaging Type:
6-Place
8-Place
No. of Places:
6
8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type No. of Places
148908 6-Place 6
148906 8-Place 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 148908 Supplier BD Supplier No. 260661
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
  • Self-tamping, complete with storage canister and reusable indicator desiccant container
  • Choose from 6-place or 8-place
Specifications
For Use With (Equipment) Sensi-Disc™ antibiotic susceptibility test disc
Includes Storage canister, reusable indicator desiccant container
No. of Places 6
Packaging Type 6-Place
Quantity Each