missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Pseudosel™ Agar (Cetrimide Agar)
For the selective isolation and presumptive identification of Pseudomonas aeruginosa from clinical and nonclinical specimens
Supplier: BD 297882
Description
- Ready-to-use
- 100 X 10mm and 100 X 15mm style plates
Compliance
Meets USP/EP/JP performance specifications.
Specifications
| ISO/DIS, 11014 | |
| 2°C to 8°C | |
| United States | |
| Pancreatic digest of Gelatin 20g/L, Potassium Sulfate 10g/L, Magnesium Chloride 1.4g/L, Agar 13.6g/L, Cetrimide 0.3g/L, Glycerol 10mL | |
| Plate | |
| 810 g |
| Cream | |
| Liquid | |
| Pseudosel agar (Cetrimide agar) | |
| Selective isolation and presumptive identification of Pseudomonas aeruginosa from clinical and nonclinical specimens | |
| 10/Pk. |