missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Phenylethyl Alcohol Agar with 5% Sheep Blood
For the isolation of gram-positive organisms, particularly gram-positive cocci, from specimens of mixed gram-positive and gram-negative flora and for determination of hemolytic reactions.
$425.68 - $425.68
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
|---|---|
| Color | Red |
| Format | Mono-Plate Format |
| Manufacturing Origin | United States |
| Media Type | Phenylethyl Alcohol Agar with 5% Sheep Blood |
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
| ISO/DIS, 11014 | |
| Mono-Plate Format | |
| Phenylethyl Alcohol Agar with 5% Sheep Blood | |
| Plate |
| Red | |
| United States | |
| Gram positive organisms, particularly gram positive cocci, from mixed specimens |