missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Media: Phenylethyl Alcohol Agar with 5% Sheep Blood

Catalog No. B21277
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21277 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21277 Supplier BD Supplier No. 221277
Only null left

For the isolation of gram-positive organisms, particularly gram-positive cocci, from specimens of mixed gram-positive and gram-negative flora and for determination of hemolytic reactions.

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Content And Storage 2°C to 8°C
Format Mono-Plate Format
Manufacturing Origin United States
Media Type Phenylethyl Alcohol Agar with 5% Sheep Blood
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, beta-Phenylethyl Alcohol 2.5g/L, Agar 15g/L, Sheep Blood, defibrinated 5%
For Use With (Application) Gram positive organisms, particularly gram positive cocci, from mixed specimens
Packaging Type Plate
Quantity 100/Pk.