missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Mueller Hinton II Agar
For antimicrobial disc diffusion susceptibility testing of common, rapidly growing bacteria by the Bauer-Kirby method
Supplier: BD 221275
Description
- Ready-to-use
- 100 x 10mm, 100 x 15mm and 150 x 15mm-style plates
Specifications
| ISO/DIS, 11014 | |
| 2°C to 8°C | |
| United States | |
| Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L | |
| 100 x 15mm Plate |
| Beige | |
| Solid | |
| Mueller Hinton II Agar | |
| Antimicrobial disc diffusion susceptibility testing of common rapidly growing bacteria by the Bauer-Kirby method | |
| 100/Pk. |