missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Mannitol Salt Agar
For the selective isolation and enumeration of staphylococci from clinical and nonclinical materials
$222.77 - $222.77
Specifications
| Color | Red |
|---|---|
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Mannitol Salt Agar |
| Catalog Number | Mfr. No. | Quantity | Certifications/Compliance | For Use With (Application) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Certifications/Compliance | For Use With (Application) | Price | Quantity | |||
|
B21271
|
BD
221271 |
100/Pk. | ISO, 9000 | Staphylococci detection/count |
Case of 100 Each for $222.77
|
|
|||
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
| Red | |
| Liquid | |
| Mannitol Salt Agar | |
| Plate |
| 2°C to 8°C | |
| United States | |
| Staphylococci detection/count |