missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Media: Mannitol Salt Agar

Catalog No. B21271
Change view
Click to view available options
Quantity:
100/Pk.
Certifications/Compliance:
ISO, 9000
For Use With (Application):
Staphylococci detection/count
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Certifications/Compliance For Use With (Application)
B21271 100/Pk. ISO, 9000 Staphylococci detection/count
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21271 Supplier BD Supplier No. 221271
Only null left

For the selective isolation and enumeration of staphylococci from clinical and nonclinical materials

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

Specifications

Certifications/Compliance ISO, 9000
Color Red
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Mannitol Salt Agar
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Beef extract 1g/L, Sodium Chloride 75g/L, D-Mannitol 10g/L, Phenol red 25mg/L, Agar 15g/L
For Use With (Application) Staphylococci detection/count
Packaging Type Plate
Quantity 100/Pk.