missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: MacConkey II Agar
Selective and differential media for the detection of coliform organisms and enteric pathogens
$179.80 - $179.80
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
|---|---|
| Color | Light Beige |
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | MacConkey II Agar |
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
| ISO/DIS, 11014 | |
| Solid | |
| MacConkey II Agar | |
| Coliform organisms and enteric pathogens |
| Light Beige | |
| United States | |
| Pancreatic digest of Gelatin 17g/L, Pancreatic digest of Casein 1.5g/L, Peptic digest of Animal Tissue 1.5g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 0.001g/L, Agar 13.5g/L | |
| Plate |