missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Hektoen Enteric Agar
For the isolation and cultivation of gram-negative enteric microorganisms, especially Shigella, from a variety of clinical and nonclinical specimens
Supplier: BD 221366
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
| ISO/DIS, 11014 | |
| Solid | |
| Hektoen enteric Agar | |
| Gram negative enteric microorganisms, especially Shigella | |
| 100/Pk. |
| Light Beige | |
| United States | |
| Peptic digest of Animal Tissue 12g/L, Yeast extract 3g/L, Bile Salts 9g/L, Lactose 12g/L, Sucrose 12g/L, Salicin 2g/L, Sodium Chloride 5g/L, Sodium Thiosulfate 5g/L, Ferric Ammonium Citrate 1.5g/L, Bromthymol blue 0.064g/L, Acid Fuchsin 0.1g/L, Agar 13.5g/L | |
| Plate | |
| 3000 g |