missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Media: Group A Selective Strep Agar with 5% Sheep Blood (ssA™)

Catalog No. B21780X
Change view
Click to view available options
Quantity:
100/Pk.
Media Type:
Selective Strep Agar, 5% SB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
B21780X 100/Pk. Selective Strep Agar, 5% SB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21780X Supplier BD Supplier No. 221780
Only null left

For the isolation of group A streptococci (S. pyogenes) from throat cultures and other specimens

This media is designed for use in conjunction with Taxo A (bacitracin, 0.04 unit) discs for presumptive identification of S. pyogenes.

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Content And Storage 2°C to 8°C
Form Solid
Manufacturing Origin United States
Media Type Selective Strep Agar, 5% SB
Formulation Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, selective Agents 40.2mg/L, Sheep Blood, defibrinated 5%
For Use With (Application) Group A streptococci (S. pyogenes) from throat cultures and other specimens
Packaging Type Plate
Quantity 100/Pk.