missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Chocolate II Agar (GC II Agar with Hemoglobin and IsoVitaleX™)
For the isolation and cultivation of fastidious microorganisms, especially Neisseria and Haemophilus species, from a variety of clinical specimens
Supplier: BD 221267
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
| ISO/DIS, 11014 | |
| 2°C to 8°C | |
| United States | |
| Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 5g/L, Agar 12g/L, Hemoglobin 10g/L, IsoVitaleX™ enrichment 10mL | |
| Plate | |
| 3800 g |
| Dark Brown | |
| Agar | |
| Chocolate II Agar (GC II Agar with Hemoglobin and IsoVitaleX™) | |
| Neisseria and Haemophilus species | |
| 100/Pk. |