missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Campy CVA Agar
Selective media used in the primary isolation and cultivation of Campylobacter jejuni from human fecal specimens
$23.75 - $68.11
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
|---|---|
| Color | Dark Red |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Campylobacter Cefoperazone Vancomycin-Amphotericin B Agar |
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
| ISO/DIS, 11014 | |
| Liquid | |
| Campylobacter Cefoperazone Vancomycin-Amphotericin B Agar | |
| Campylobacter jejuni from Human feces |
| Dark Red | |
| United States | |
| Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 15g/L, Cefoperazone 20mg/L, Vancomycin 10mg/L, Amphotericin B 2mg/L, Sheep Blood, defibrinated | |
| Plate |