missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Media: BCYE Selective Agar

Catalog No. p-4766447
Change view
Click to view available options
Color:
Dark Green
Media Type:
BCYE selective Agar with PAC
Format:
Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Media Type Format
B97879X Dark Green BCYE selective Agar with PAC Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B97879X Supplier BD Supplier No. 297879
Edge

For isolation of Legionella species from clinical and nonclinical specimens

  • Enables the isolation of microorganisms from samples or specimens.
  • Ready for immediate use in Petri-style dishes containing a variety of media.

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Green
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Polymyxin B, Anisomycin, Cefamandole
Detects Legionella species
Form Liquid
Format Plate
Manufacturing Origin United States
Media Type BCYE selective Agar with PAC
Product Type Prepared Microbiology Plate
Test Type Media Testing
Formulation BCYE Agar 38.65g/L, Polymyxin B 40000U, Anisomycin 80mg/L, Cefamandole 4mg/L
For Use With (Application) Legionella species from contaminated clinical specimens (with cefamandole)
Packaging Type Plate
Quantity 10/Pk.
Show More Show Less