missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Media: BCYE Agar (for Legionella)

Catalog No. B21808
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21808 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21808 Supplier BD Supplier No. 221808
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For primary isolation and cultivation of Legionella pneumophila and other Legionella species from environmental samples and clinical specimens

  • For primary isolation and cultivation of Legionella pneumophila and other Legionella species from environmental samples and clinical specimens.

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Black
Country of Origin United States
Additive L-cysteine HCl, Ferric Pyrophosphate, ACES Buffer, Charcoal, Activated, Alpha-ketoglutarate
Detects Legionella pneumophila, Legionella species
Form Liquid
Format Prepared Agar Plate
Manufacturing Origin United States
Media Type BCYE Agar (for Legionella)
Product Type Prepared Microbiology Plate
Target Organism Class Bacteria
Test Type Microbial Cultivation
Formulation Yeast extract 10g/L, L-Cysteine HCl 0.4g/L, Ferric Pyrophosphate 0.25g/L, ACES Buffer 10g/L, Charcoal, Activated 2g/L, Alpha-Ketoglutarate 1g/L, Agar 15g/L
For Use With (Application) Primary isolation and cultivation of Legionella pneumophila and other Legionella species from environmental samples and clinical specimens
Packaging Type Plate
Quantity 10/Pk.
Manufacturing Quality cGMP Standards, FDA-inspected, ISO 9001 Certified
Show More Show Less