missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Chromogenic Media: m-EI Agar Base
Description
- Any colonies present will exhibit blue halo within 24 hours of the test
- Conforms to EPA Method 1600
Order Info
Available in prepared 15 x 60mm plates.
Spécifications
Spécifications
| Certifications/Compliance | US, EPA |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | m EI Agar |
| Formulation | Peptone 10g/L, Sodium Chloride 15g/L, Esculin 1g/L, Cycloheximide 0.05g/L, Sodium Azide 0.15g/L, Yeast extract 30g/L, Indoxyl-beta-D-glucoside 0.75g/L, Agar 15g/L, Nalidixic Acid 0.24g/L, Triphenyltetrazolium Chloride 0.02g/L |
| For Use With (Application) | The chromogenic detection and enumeration of Enterococci in Water by the single-step membrane filtration technique |
| Packaging Type | Plate |
| Quantity | 100/Pk. |