missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Media Mossel Enterobacteriaceae Enrichment Broth
For isolation of bile-tolerant gram-negative bacteria from nonsterile pharmaceutical products
Supplier: BD 292627
Description
- Prepared Bottled Media, ready for immediate use
- Wide mouth bottle
- Used for selectively enriching and detecting Enterobacteriaceae in food
- With transparent labeling to allow clear view of media and culture
- Meets United States Pharmacopeia (USP), European Pharmacopoeia (EP) and Japanese Pharmacopoeia (JP)1-3 performance specifications
- Appearance: Medium to dark green and clear to trace hazy
- Reaction at 25°C, pH: 7.2 ± 0.2
- Volume: 90mL, Packaging: 10 Each
Specifications
| ISO/DIS, 11014, COMPF, EP, ISO, JP, USP | |
| Liquid | |
| Mossel Enterobacteriaceae Enrichment Broth | |
| Detection of Enterobacteriaceae, type: Bottle, wide mouth | |
| 90 mL |
| 2°C to 8°C | |
| United States | |
| Pancreatic digest of Gelatin 10g/L, Dextrose 5g/L, Oxgall 20g/L, Disodium Phosphate 8g/L, Monopotassium Phosphate 2g/L, Brilliant green 15mg/L | |
| Tube with Tight Cap |