missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Bottled Media: Buffered Sodium Chloride Peptone Solution (pH 7.0)

Used for dissolving, suspending and diluting test samples

$644.50 - $644.50

Specifications

Color Clear
Content And Storage 5°C to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Buffered Sodium Chloride Peptone solution
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Certifications/Compliance Packaging Type Price Quantity  
Catalog Number Mfr. No. Quantity Certifications/Compliance Packaging Type Price Quantity  
B257086
View Documents
BD
257086
100 mL ISO/DIS, 11014 Sterile prepared infusion bottle, 25/Pk
Pack of 25 Each for $644.50
Only null left
 
Description

Description

  • Used to make suspensions of organisms for testing growth promoting and inhibitory properties of media when examining non-sterile pharmaceutical products for specified microorganisms
  • Fluid provides osmotic stability, stable pH value
  • Maintains viability of microorganisms during preparation of samples
  • Surface-active ingredients or inactivators of antimicrobial agents
Specifications

Specifications

Clear
Liquid
Buffered Sodium Chloride Peptone solution
Injection stopper and pull ring
5°C to 25°C
United States
Detection of micororganisms from pharmaceutical, food, environmental or Water testing samples
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'