missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Mycoflask™ Thioglycollate Prepared Media Enriched with Vitamin K1 and Hemin

Catalog No. L21742
Change view
Click to view available options
Quantity:
100 x 5 mL
100 x 8 mL
100 x 10 mL
Color:
Beige
Light Yellow
Packaging Type:
Size A tube with tight cap
Size K tube with tight cap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color Packaging Type
L21742 100 x 5 mL Beige Size K tube with tight cap
B4321788 100 x 8 mL Light Yellow Size K tube with tight cap
L97292 100 x 10 mL Beige Size A tube with tight cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L21742 Supplier BD Supplier No. 221742
Only null left

Ideal for use in the isolation and cultivation of fastidious or slow growing, obligately anaerobic microorganisms present in clinical materials

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

Specifications

Certifications/Compliance CE, BS12, CLSI, CMPH2, MCM9
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Thioglycollate Medium
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.7g/L, L-Cystine 0.25g/L, Sodium Sulfite 0.1g/L, Hemin 0.005g/L, Vitamin K1 0.001g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms
Packaging Type Size K tube with tight cap
Quantity 100 x 5 mL

For in vitro diagnostic use.