missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Mycoflask™ Prepared Pour Tubed Media: Trypticase™ Soy Agar (Blood Agar Base) Deeps

Catalog No. 221082
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
221082 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 221082 Supplier BD Supplier No. 221082
Only null left

For the isolation and cultivation of fastidious and nonfastidious anaerobic and aerobic microorganisms

  • For cultures requiring prolonged incubation

Specifications

Certifications/Compliance CE, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Color Beige
Form Liquid
Manufacturing Origin United States
Media Type Trypticase soy agar
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Isolation and cultivation of non-fastidious and fastidious microorganisms
Packaging Type Tube
Quantity 20 mL