Learn More
BD BBL™ Mycoflask™ Nutrient Broth Prepared Media
For cultivation of many species of nonfastidious microorganisms
Supplier:  BD 221669
Catalog No. B21669
Only null left
Explore available promotions
Description
Description
Nutrient Broth has the formula originally designed for use in the standard methods for the examination of water and wastewater.
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
CE, BAM, CCAM, COMPF, SMD
Powder
Nutrient Broth
Cultivation of many species of non-fastidious microorganisms
5 mL
Beige
United States
Pancreatic digest of Gelatin 5g/L, Beef extract 3g/L
Tube with Tight Cap
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For in vitro diagnostic use.