missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ GN (Gram-Negative) Broth, 8 mL
Used for selective enrichment of Salmonella and Shigella
Supplier: BD 221729
Requires : Ancillary culture media, reagents, quality control organisms and laboratory equipment
Description
Inoculate broth as soon as possible after specimens arrive at laboratory.
Specifications
| CE, BS12, CCAM, CMPH2, COMPF, MCM9 | |
| 2°C to 8°C | |
| United States | |
| Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, D-Mannitol 2g/L, Sodium Citrate 5g/L, Sodium Desoxycholate 0.5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L | |
| 10 | |
| 8 mL |
| Light Beige | |
| Liquid | |
| GN Broth | |
| Selective enrichment of Salmonella and Shigella | |
| Tube |
For in vitro diagnostic use.