missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™/Difco™ TB Stain Kit K

Catalog No. B4312522
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4312522 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4312522 Supplier BD Supplier No. 212522
Only null left

Includes

One 250mL bottle each: Carbolfuchsin KF, Decolorizer and Brilliant Green K

For staining mycobacteria by the Kinyoun (cold) acid-fast procedure

Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.

Specifications

Type TB Kinyoun Stain Kit
Quantity 1 Ea.
Includes Carbol fuchsin KF, decolorizer, and brilliant green K
Format Bottle
For Use With (Application) For staining mycobacteria by the Kinyoun (cold) acid-fast procedure
Storage Requirements 15° to 30°C
Packaging Type Bottle
Stain Type Stain Set K (Carbolfuchsin KF, Decolorizer and Brilliant Green K)

For in vitro Diagnostic Use.