Learn More
BD BBL™/Difco™ TB Methylene Blue
For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures
Supplier: BD B212516
Description
Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.
Specifications
| TB Methylene Blue | |
| For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures | |
| Bottle |
| 4 x 250 mL | |
| 15° to 30°C | |
| Methylene Blue |
For in vitro Diagnostic Use.