missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dermatophyte Test Medium (DTM) Modified with Chloramphenicol Slants
Description
Requires
Ancillary culture media, reagents, quality control organisms and laboratory equipment
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
| Certifications/Compliance | CE, BS12, MCM9 |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Dermatophyte Test Medium |
| Formulation | Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 24g/L, Phenol red 0.2g/L, Cycloheximide 0.5g/L, gentamicin 0.1g/L, Chloramphenicol 0.1g/L |
| For Use With (Application) | Detection and presumptive identification of dermatophytes |
| Packaging Type | Tube |
| Quantity | 10/Pk. |
For in vitro diagnostic use.