missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Yeast Extract Agar

Catalog No. B11546
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11546 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11546 Supplier BD Supplier No. 211546
Edge

For isolation of Dermatophytes, Trichophyton verrucosum

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 25°C
Form Dehydrated
Media Type Yeast Extract Agar
Product Type Agar
Formulation Papaic digest of Soybean Meal 10g/L, Yeast extract 5g/L, Dextrose 40g/L, Streptomycin 0.03g/L, Chloramphenicol 0.05g/L, Agar 17g/L
For Use With (Application) Isolation of Dermatophytes, Trichophyton verrucosum
Packaging Type Bottle
Quantity 500 g