missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Trypticase Soy Agar (TSA II)
Description
Supplementad with blood is used for cultivating fastidious microorganisms and for hemolytic reactions produced by many bacterial species
Certifications
BAM, COMPF
Specifications
Specifications
| Certifications/Compliance | BAM, COMPF |
| Content And Storage | 2 to 25°C |
| Form | Dehydrated |
| Media Type | Trypticase Soy Agar (TSA II) |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L |
| For Use With (Application) | Cultivation of fastidious microorganisms and For the visualization of hemolytic reactions |
| Packaging Type | Tube |
| Quantity | 500 g |