missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Soybean Casein Digest Agar
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
$1162.37 - $4883.39
Specifications
| Certifications/Compliance | ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP |
|---|---|
| Content And Storage | 25°C |
| Form | Dehydrated |
| Media Type | Soy Agar |
| Product Type | Agar |
Description
Applications:
Isolation of non-fastidious and fastidious
Specifications
| ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP | |
| Dehydrated | |
| Agar | |
| Isolation of non-fastidious and fastidious |
| 25°C | |
| Soy Agar | |
| Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L | |
| Pail |