missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Mycosel™ Agar
Specifications
Specifications
| Certifications/Compliance | BS12, CMPH2, MCM9 |
| Form | Dehydrated |
| Media Type | Mycosel Agar |
| Product Type | Agar |
| Formulation | Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 15.5g/L, Cycloheximide 0.4g/L, Chloramphenicol 0.05g/L |
| For Use With (Application) | Isolation of pathogenic fungi from materials having a large flora of other fungi and bacteria |
| Packaging Type | Bottle |
| Quantity | 500 g |