missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Mycoplasma Broth Base (Frey)
For the cultivation of avian mycoplasmas
Supplier: BD 212346
Specifications
| Dehydrated | |
| Microbiology Media | |
| Cultivation of avian mycoplasmas | |
| 500 g |
| Mycoplasma Broth Base | |
| Pancreatic digest of Casein 7.5g/L, Papaic digest of Soybean Meal 2.5g/L, Yeast extract 5g/L, Sodium Chloride 5g/L, Potassium Chloride 0.4g/L, Magnesium Sulfate 0.2g/L, Disodium Phosphate 1.6g/L, Monopotassium Phosphate 0.1g/L | |
| Bottle |