missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Mueller Hinton II Broth (Cation-Adjusted)

Catalog No. B12322
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12322 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B12322 Supplier BD Supplier No. 212322
Edge

For antimicrobial susceptibility testing of microorganisms by broth dilution methods. BD Difco™ BBL™ Dehydrated Culture Media: Mueller Hinton II Broth (Cation-Adjusted) is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Mueller Hinton II Broth (Cation-Adjusted)

Specifications

Certifications/Compliance BS12, CLSI, CMPH2, MCM9
Content And Storage 2 to 25°C
Form Dehydrated
Media Type Mueller Hinton II Broth
Product Type Microbiology Media
Formulation Beef extract 3g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L
For Use With (Application) Antimicrobial susceptibility testing of microorganisms by Broth dilution method
Packaging Type Bottle
Quantity 500 g