missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Mannitol Salt Agar

Catalog No. B11407
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11407 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11407 Supplier BD Supplier No. 211407
Edge

For isolation of pathogenic staphylococci

Certifications

BAM, BS12, CMPH2, EP, JP, MCM9, USP

Specifications

Certifications/Compliance BAM, BS12, CMPH2, EP, JP, MCM9, USP
Form Dehydrated
Media Type Mannitol Salt Agar
Product Type Mannitol Salt Agar
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Beef extract 1g/L, Sodium Chloride 75g/L, D-Mannitol 10g/L, Phenol red 25mg/L, Agar 15g/L
For Use With (Application) For selective isolation and enumeration of staphylococci from clinical and nonclinical materials
Packaging Type Bottle
Quantity 500 g